missing translation for 'onlineSavingsMsg'
Learn More

Strontium peroxide, 12.3% available oxygen

Catalog No. p-7080070
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4631030 250 g
AA46310A1 1 kg
AA46310A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4631030 Supplier Thermo Scientific Chemicals Supplier No. 04631030
Only null left

CAS: 1314-18-7 | O2Sr | 119.618 g/mol

Applications of Strontium Peroxide include pyrotechnics i.e. fireworks (red color), ammunitions (tracer/hunting), oxidizer, alarm rocket. It is also used in textiles for fibers bleaching and as an oxidizing agent for hair bleaching (professional).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-18-7
Molecular Formula O2Sr
Molecular Weight (g/mol) 119.618
MDL Number MFCD00084686
InChI Key UHCGLDSRFKGERO-UHFFFAOYSA-N
Synonym strontium peroxide, 1,2-dioxa-3-strontacyclopropane, strontium dioxide, strontiumperoxid, strontium peroxide sr o2, hsdb 788, strontium peroxide un1509 oxidizer, strontium peroxide available oxygen
PubChem CID 14807
IUPAC Name strontium;peroxide
SMILES [O-][O-].[Sr+2]

Specifications

Melting Point 215°C (decomposition)
Color Brown
Physical Form Powder
Quantity 250 g
Linear Formula SrO2
UN Number UN1509
Merck Index 14,8847
Solubility Information Insoluble in water and slowly decomposes in water.
Formula Weight 119.62
Sensitivity Moisture sensitive
Chemical Name or Material Strontium peroxide, 12.3% available oxygen
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H272-H318-H335-H315
May intensify fire
oxidizer.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 5.1
EINECSNumber 215-224-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only