missing translation for 'onlineSavingsMsg'
Learn More

Strontium hydroxide octahydrate, 98%, pure

Catalog No. AC270230010
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270230010 1 kg Glass bottle
AC270230050 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC270230010 Supplier Thermo Scientific Chemicals Supplier No. 270230010

CAS: 1311-10-0 | H18O10Sr | 265.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1311-10-0
Molecular Formula H18O10Sr
Molecular Weight (g/mol) 265.75
MDL Number MFCD00150395
InChI Key UJPWWRPNIRRCPJ-UHFFFAOYSA-L
IUPAC Name strontium(2+) octahydrate dihydroxide
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Sr++]

Specifications

Melting Point 100.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 98%
Linear Formula Sr(OH)2·8H2O
Merck Index 14,8843
Solubility Information Solubility in water: 7.0g/L (20°C). Other solubilities: soluble in hot water, soluble in acids
Formula Weight 265.75
Percent Purity 98%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Strontium hydroxide octahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only