missing translation for 'onlineSavingsMsg'
Learn More

Strontium granules, 19mm (0.76in) & down, 99% (metals basis)

Catalog No. AA3578906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3578906 5 g
AA3578914 25 g
AA3578922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3578906 Supplier Thermo Scientific Chemicals Supplier No. 03578906

Strontium granules, 99%, Sr, CAS Number-7440-24-6, atom, granules, 19mm 0.76in & down metals basis, estroncio, strontium, chunks, pieces, dendritic, pieces, yzs2rpe8le, strontium, elemental, strontium, distilled dendritic pieces, unii-yzs2rpe8le, 100g, (metals basis), CHEBI:33324, White to Yellow | CAS: 7440-24-6 | Sr | 87.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon with thin oil layer

Chemical Identifiers

CAS 7440-24-6
Molecular Formula Sr
Molecular Weight (g/mol) 87.62
MDL Number MFCD00134060
InChI Key CIOAGBVUUVVLOB-UHFFFAOYSA-N
Synonym strontium, elemental, unii-yzs2rpe8le, yzs2rpe8le, estroncio, granules, 19mm 0.76in & down metals basis, strontium, distilled dendritic pieces, atom, pieces, strontium, chunks, pieces, dendritic
PubChem CID 5359327
ChEBI CHEBI:33324
IUPAC Name strontium
SMILES [Sr]

Specifications

Color White to Yellow
Quantity 5 g
Particle Size 19mm (0.76 in.) & down
Sensitivity Air and moisture sensitive
UN Number UN3208
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Strontium granules
Hazard Category H260
Hazard Statement GHS H Statement
H261
In contact with water releases flammable gas.
Precautionary Statement P223-P231+P232-P280-P335+P334-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
EINECSNumber 231-133-4
RTECSNumber WK7700000
TSCA Yes
storageNote1
  • Packaged under argon with thin oil layer
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only