Learn More
Strontium fluoride, 99% min
CAS: 7783-48-4 | F2Sr | 125.62 g/mol
$106.08 - $1082.14
Chemical Identifiers
| CAS | 7783-48-4 |
|---|---|
| Molecular Formula | F2Sr |
| Molecular Weight (g/mol) | 125.62 |
| MDL Number | MFCD00011251 |
| InChI Key | FVRNDBHWWSPNOM-UHFFFAOYSA-L |
| Synonym | strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g |
| PubChem CID | 82210 |
| IUPAC Name | strontium;difluoride |
| SMILES | [F-].[F-].[Sr++] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1220130
|
Thermo Scientific Chemicals
01220130 |
250 g |
Each for $106.08
|
|
|||||
|
AA12201A1
|
Thermo Scientific Chemicals
012201A1 |
1 kg |
Each for $271.86
|
|
|||||
|
AA12201A7
|
Thermo Scientific Chemicals
012201A7 |
5 kg |
Each for $1,082.14
|
|
|||||
Description
Strontium fluoride is used as an optical material for a small range of special applications. It also used as use is as a carrier of strontium-90 radioisotope in radioisotope thermoelectric generators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7783-48-4 | |
| 125.62 | |
| FVRNDBHWWSPNOM-UHFFFAOYSA-L | |
| 82210 | |
| [F-].[F-].[Sr++] |
| F2Sr | |
| MFCD00011251 | |
| strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g | |
| strontium;difluoride |
Specifications
| 1473°C | |
| White | |
| 250 g | |
| MFCD00011251 | |
| strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g | |
| FVRNDBHWWSPNOM-UHFFFAOYSA-L | |
| strontium;difluoride | |
| 82210 | |
| ≤99% | |
| 1.442 | |
| Strontium fluoride |
| 7783-48-4 | |
| Powder | |
| F2Sr | |
| 14,8842 | |
| Slightly soluble in water (11;7mg at 18°C/100mL), dilute acids; Decomposes in strong acids | |
| [F-].[F-].[Sr++] | |
| 125.62 | |
| 125.62 | |
| Odorless | |
| 4.240 g/mL |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 232-000-3
RTECSNumber : WK8925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only