missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium chloride hexahydrate, Puratronic™, 99.9965% (metals basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 5 g |
| Merck Index | 14,8840 |
| Formula Weight | 266.62 (158.53 Anhydrous) |
| Percent Purity | 99.9965% |
| Grade | Puratronic |
| Refractive Index | 1.65 |
| Sensitivity | Hygroscopic |
| Density | 1.93 g/mL |
| Show More |
Safety and Handling
| Hazard Category | H318-H335 |
| Hazard Statement | GHS H Statement H318 Causes serious eye damage. |
| Precautionary Statement | P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c |
| EINECSNumber | 233-971-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only