missing translation for 'onlineSavingsMsg'
Learn More

Strontium chloride hexahydrate, ACS, 99.0-103.0%

Catalog No. p-7050029
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3339322 100 g
AA3339336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3339322 Supplier Thermo Scientific Chemicals Supplier No. 03339322
Only null left

CAS: 10025-70-4 | Cl2H12O6Sr | 266.61 g/mol

Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the medical field, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10025-70-4
Molecular Formula Cl2H12O6Sr
Molecular Weight (g/mol) 266.61
MDL Number MFCD00149865
InChI Key AMGRXJSJSONEEG-UHFFFAOYSA-L
IUPAC Name strontium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]

Specifications

Physical Form Granules
Quantity 100 g
Assay Percent Range 99.0 to 103.0%
Merck Index 14,8840
Solubility Information Soluble in water. Slightly soluble in ethanol and acetone. Insoluble in ammonia.
Formula Weight 266.62 (158.53 Anhydrous)
Percent Purity 99.0 to 103.0%
Grade ACS
Refractive Index 1.65
Sensitivity Hygroscopic
Density 1.93 g/mL
Chemical Name or Material Strontium chloride hexahydrate, For analysis ACS
Show More Show Less
Hazard Category H318-H335
Hazard Statement GHS H Statement
H318
Precautionary Statement P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
EINECSNumber 233-971-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only