missing translation for 'onlineSavingsMsg'
Learn More

Strontium chloride hexahydrate, 98%

CAS: 10025-70-4 | Cl2H12O6Sr | 266.61 g/mol

Supplier:  Thermo Scientific Chemicals A167640B

 View more versions of this product

Catalog No. AAA167640B


Only null left
Explore available promotions

Description

Description

Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the medical field, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the medical field, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.

Solubility
Soluble in water. Slightly soluble in ethanol and acetone. Insoluble in ammonia.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Strontium chloride hexahydrate
1000 g
MFCD00149865
Soluble in water. Slightly soluble in ethanol and acetone. Insoluble in ammonia.
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
266.61
≥99%
Hygroscopic
10025-70-4
Cl2H12O6Sr
14,8840
AMGRXJSJSONEEG-UHFFFAOYSA-L
strontium(2+) hexahydrate dichloride
266.62 (158.53 Anhydrous)
1.65
1.93

Chemical Identifiers

10025-70-4
266.61
AMGRXJSJSONEEG-UHFFFAOYSA-L
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
Cl2H12O6Sr
MFCD00149865
strontium(2+) hexahydrate dichloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c

H318-H335

EINECSNumber : 233-971-6

RTECSNumber : WK8575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only