Learn More
Strontium bromide hexahydrate, 95%
CAS: 7789-53-9 | Br2H12O6Sr | 355.518 g/mol
$79.60 - $250.67
Chemical Identifiers
| CAS | 7789-53-9 |
|---|---|
| Molecular Formula | Br2H12O6Sr |
| Molecular Weight (g/mol) | 355.518 |
| MDL Number | MFCD00150361 |
| InChI Key | FLMJUJXBFKFYOZ-UHFFFAOYSA-L |
| Synonym | unii-7i32n2ud6w, strontium dibromide hexahydrate, strontium bromide, reagent, strontium 2+ hexahydrate dibromide, strontium 2+ ion hexahydrate dibromide |
| PubChem CID | 165644 |
| IUPAC Name | strontium;dibromide;hexahydrate |
| SMILES | O.O.O.O.O.O.[Br-].[Br-].[Sr+2] |
Description
Strontium bromide hexahydrate is used in flares and also used in some pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7789-53-9 | |
| 355.518 | |
| FLMJUJXBFKFYOZ-UHFFFAOYSA-L | |
| 165644 | |
| O.O.O.O.O.O.[Br-].[Br-].[Sr+2] |
| Br2H12O6Sr | |
| MFCD00150361 | |
| unii-7i32n2ud6w, strontium dibromide hexahydrate, strontium bromide, reagent, strontium 2+ hexahydrate dibromide, strontium 2+ ion hexahydrate dibromide | |
| strontium;dibromide;hexahydrate |
Specifications
| 7789-53-9 | |
| 100 g | |
| Br2H12O6Sr | |
| unii-7i32n2ud6w, strontium dibromide hexahydrate, strontium bromide, reagent, strontium 2+ hexahydrate dibromide, strontium 2+ ion hexahydrate dibromide | |
| FLMJUJXBFKFYOZ-UHFFFAOYSA-L | |
| strontium;dibromide;hexahydrate | |
| 165644 | |
| Hygroscopic | |
| Strontium bromide hexahydrate |
| Crystalline | |
| 95% | |
| MFCD00150361 | |
| Soluble in water,alcohol | |
| O.O.O.O.O.O.[Br-].[Br-].[Sr+2] | |
| 355.518 | |
| 355.53 (247.44 Anhydrous) | |
| 2.386 g/mL |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 233-969-5
RTECSNumber : WK8225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only