missing translation for 'onlineSavingsMsg'
Learn More

Strontium bis(trifluoromethylsulfonyl)imide

Catalog No. p-7052252
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4709903 1 g
AA4709906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4709903 Supplier Thermo Scientific Chemicals Supplier No. 04709903
Only null left

CAS: 862121-57-1 | C4F12N2O8S4Sr | 647.891 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 862121-57-1
Molecular Formula C4F12N2O8S4Sr
Molecular Weight (g/mol) 647.891
MDL Number MFCD23380176
InChI Key DIMYICIXZTVMOY-UHFFFAOYSA-N
Synonym Bis(trifluoromethane)sulfonimide strontium salt; Bis(trifluoromethanesulfonyl)imide strontium salt
PubChem CID 134159298
IUPAC Name strontium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Sr+2]

Specifications

Melting Point >200°C
Color White
Physical Form Solid
Quantity 1 g
Linear Formula [(CF3SO2)2N]2Sr
UN Number UN3261
Formula Weight 647.91
Sensitivity Hygroscopic
Chemical Name or Material Strontium bis(trifluoromethylsulfonyl)imide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only