missing translation for 'onlineSavingsMsg'
Learn More

Strontium aluminate, 99.5% (metals basis)

Catalog No. AA3964722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3964722 100 g
AA3964714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3964722 Supplier Thermo Scientific Chemicals Supplier No. 03964722
Only null left

CAS: 12004-37-4 | SrAl2O4

Strontium aluminate acts as a photo luminescent phosphor with long persistence of phosphorescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12004-37-4
Molecular Formula SrAl2O4
MDL Number MFCD00054054
Synonym strontium aluminate, strontium;oxido oxo alumane, dioxo-2,4-dioxa-1,5-dialumina-3-strontapentane, strontium aluminate,-100 mesh trace metals basis

Specifications

Physical Form Powder
Quantity 100 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Slightly soluble in water.
Formula Weight 205.58
Density 3.559
Chemical Name or Material Strontium aluminate
EINECSNumber 234-455-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only