missing translation for 'onlineSavingsMsg'
Learn More

Strontium acetate hemihydrate, Reagent Grade

Catalog No. p-7048959
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1220330 250 g
AA12203A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1220330 Supplier Thermo Scientific Chemicals Supplier No. 01220330
Only null left

CAS: 14692-29-6

Strontium acetate is used as a catalyst, as a chemical intermediate and in medicine. The primary application of strontium acetate is as an active component in toothpastes. This compound is used to desensitize teeth to extremes of hot and cold, sweets, acids, or touch.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14692-29-6
MDL Number MFCD00198064

Specifications

Melting Point 150°C (in -0.5H2O)
Physical Form Crystalline
Quantity 250 g
Merck Index 14,8836
Solubility Information Soluble in water and slightly soluble in alcohol.
Formula Weight 214.72 (205.71 Anhydrous)
Grade Reagent
Density 2.099
Chemical Name or Material Strontium acetate hemihydrate
EINECSNumber 208-854-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only