missing translation for 'onlineSavingsMsg'
Learn More

Strontium acetate hemihydrate, 98%

CAS: 14692-29-6 | Sr(OOCCH3)2·0.5H2O

Supplier:  Thermo Scientific Chemicals A161490E

 View more versions of this product

Catalog No. AAA161490E


Only null left
Explore available promotions

Description

Description

PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Strontium acetate hemihydrate
150°C (in -0.5H2O)
MFCD00198064
14,8836
Soluble in water.
98%
14692-29-6
Sr(OOCCH3)2·0.5H2O
2500 g
Acetic acid strontium salt
214.72 (205.71 Anhydrous)

Chemical Identifiers

14692-29-6
Sr(OOCCH3)2·0.5H2O
MFCD00198064
Acetic acid strontium salt
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-854-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only