missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Strontium

Catalog No. 44189925G
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44189925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44189925G Supplier Sigma Aldrich Supplier No. 44189925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Strontium

Specifications

Melting Point 757°C
CAS 7440-24-6
Boiling Point 1384°C
Density 2.6 g/mL (at 25°C)
Quantity 25 g
Molecular Formula Sr
Linear Formula Sr
MDL Number MFCD00134060
Synonym Strontium element
Molecular Weight (g/mol) 87.62
Percent Purity 99.99% trace metals basis
Show More Show Less