missing translation for 'onlineSavingsMsg'
Learn More

Strontium 2-ethylhexanoate, 40% in 2-ethylhexanoic acid (8-12% Sr)

CAS: 2457-02-5 | C6H8O4 | 144.13 g/mol

$300.10 - $1179.22

Chemical Identifiers

CAS 2457-02-5
MDL Number MFCD00070438
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4071914
View Documents
Thermo Scientific Chemicals
04071914
25 g
Each for $300.10
Only null left
 
AA4071922
View Documents
Thermo Scientific Chemicals
04071922
100 g
Each for $1,179.22
Only null left
 
Description

Description

Strontium 2-ethylhexanoate is used in metal refining, plumbing, bleaching, engraving, plating, photography, disinfection, fertilizer manufacture, metal cleaning, and rust removal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2457-02-5
MFCD00070438

Specifications

2457-02-5
C16H30O4Sr
25 g
Insoluble in water.
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sr+2]
374.03
374.04
Liquid
Mild
MFCD00070438
strontium 2-ethylhexanoate, strontium bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, strontium salt 2:1, acmc-20ajmv, bis 2-ethylhexanoic acid strontium salt, hexanoic acid, 2-ethyl-, strontium salt, strontium 2+ bis 2-ethylhexanoate, hexanoic acid,2-ethyl-, strontium salt 2:1
DLWBHRIWCMOQKI-UHFFFAOYNA-L
strontium;2-ethylhexanoate
102822
40% in 2-ethylhexanoic acid (8 to 12% Sr)
Strontium 2-ethylhexanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H361d

EINECSNumber : 219-536-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only