missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium 2-ethylhexanoate, 40% in 2-ethylhexanoic acid (8-12% Sr)
Description
Strontium 2-ethylhexanoate is used in metal refining, plumbing, bleaching, engraving, plating, photography, disinfection, fertilizer manufacture, metal cleaning, and rust removal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Odor | Mild |
| Molecular Formula | C6H8O4 |
| Quantity | 100 g |
| Synonym | monoethyl fumarate, e-4-ethoxy-4-oxobut-2-enoic acid, ethyl hydrogen fumarate, fumaric acid monoethyl ester, e-4-ethoxy-4-oxo-2-butenoic acid, unii-y3w849ng2n, 2e-4-ethoxy-4-oxobut-2-enoic acid, fumaric acid, monoethyl ester, mono-ethyl fumarate, ethyl hydrogen maleate |
| Solubility Information | Insoluble in water. |
| InChI Key | XLYMOEINVGRTEX-ONEGZZNKSA-N |
| SMILES | CCOC(=O)C=CC(=O)O |
| IUPAC Name | (E)-4-ethoxy-4-oxobut-2-enoic acid |
| Molecular Weight (g/mol) | 144.13 |
| PubChem CID | 5358902 |
| Show More |
Safety and Handling
| Hazard Category | H361d |
| Hazard Statement | GHS H Statement H361 Suspected of damaging fertility or the unborn child. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 219-536-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only