missing translation for 'onlineSavingsMsg'
Learn More

Strong Ammonia Solution, NF, 27-31%, Spectrum™ Chemical

Catalog No. p-7158319
Change view
Click to view available options
Quantity:
2.5 L
4 L
6 x 500 mL
200 L
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607027 2.5 L Poly-Coated Glass Bottle
18607026 6 x 500 mL Amber Glass Bottle
18607028 4 L Poly-Coated Glass Bottle
18607032 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18607027 Supplier Spectrum Chemical Mfg Cor Supplier No. AM1802.5LTPV
Only null left

AM180, 1336-21-6, NH3(aq)

Spectrum™ Chemical Strong Ammonia Solution, NF has many uses such as in topical pharmaceutical solutions destined for treating insect bites. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
IUPAC Name amine hydrate
SMILES N.O

Specifications

CAS Max % 1
Quantity 2.5 L
Percent Purity 27 to 31%
Grade NF
Packaging Poly-Coated Glass Bottle