Learn More
Corning™ Stripwell™ Microplates

Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Microplates provide a superior solution for in vitro diagnostic assays.
Supplier: Corning™ 2481
Description
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
- Stripwell microplates have 96-well flat bottom polystyrene format.
- Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
- 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Order Info
Bulk packed. Cat. No. 07-200-24 has individual 1 x 8 strips without frame.
Certifications
Cat. Nos. 07-200-31 and -32 have certified surface chemistry.
Chemical Identifiers
| Strip Plate | |
| 8 x 12 | |
| 96 | |
| In-vitro diagnostic assays | |
| 25/Pk. |
| Without Lid | |
| Polystyrene | |
| Non-sterile | |
| 100/Cs. | |
| 100/Cs. |
Specifications
| High Binding | |
| Without Lid | |
| High Bind | |
| Polystyrene | |
| 75 to 125 μL | |
| High Binding | |
| Round | |
| 100/Cs. | |
| 100/Cs. |
| Strip Plate | |
| 8 x 12 | |
| Clear | |
| 96 | |
| Non-sterile | |
| 190 μL | |
| In-vitro diagnostic assays | |
| 25/Pk. |