missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Stripettor™ Pro Pipet Controller
GREENER_CHOICE

Catalog No. 01001023
Change view
Click to view available options
Volume (Metric):
0.5 to 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01001023 0.5 to 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001023 Supplier Corning™ Supplier No. 4999
  • ONLINE EXCLUSIVE PRICE
    $801.55
    $706.35 / Case

    Save $95.20

    12.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pipet Controller with LCD display for continuous visualization of speed, modes, and battery status.

This light, ergonomic, motorized pipet controller is designed for use with glass and plastic serological pipets from 0.5 to 100 mL. Conveniently positioned switches change operating modes and speeds quickly to handle different liquid volumes and various viscosities. Aspirating and dispensing speed is controlled through the finger triggers. When fully charged, the batteries enable up to 8 hours of continuous use. The large LCD display clearly indicates battery status, pipetting mode, and speed.

  • LCD display for continuous visualization of speed, modes, and battery status
  • Multiple speed options for both aspiration and dispensing
  • Well positioned function buttons
  • 3-position charging stand
  • Lithium ion battery

Corning Stripettor Pro Pipet Controller comes with a universal power supply, 0.2 μm filter, a wall hanger, and a charging stand.

Specifications

Autoclavable Not Autoclavable
Color White
Disposable Reusable
For Use With Serological Pipettes
Grip Type Ergonomic
Material Plastic
Model Stripettor
Number of Channels 1
Sterility Non-sterile
Type Pipette Controller
Volume (Metric) 0.5 to 100 mL
Show More Show Less