missing translation for 'onlineSavingsMsg'
Learn More

Streptomycin Sulfate 95.0+%, TCI America™

Catalog No. S058525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S058525G 25 g
S0585500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S058525G Supplier TCI America Supplier No. S058525G

Chemical Identifiers

CAS 3810-74-0
Molecular Formula C42H84N14O36S3
Molecular Weight (g/mol) 1457.376
MDL Number MFCD00037023
InChI Key QTENRWWVYAAPBI-FFCQDDOVSA-N
PubChem CID 124080941
IUPAC Name 2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(
SMILES CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Type Streptomycin Sulfate
Physical Form Crystalline Powder
Color White/Yellow
Quantity 25 g
Formula Weight 728.69
Percent Purity ≥95.0% (N,T)
Chemical Name or Material Streptomycin Sulfate
RTECSNumber WK4990000
TSCA Yes