Learn More
ACROBiosystems Inc Streptavidin Coated Plates, Clear, 96-Well, Clear Frame 5 plates
Catalog No. 502330886
Encompass
This Streptavidin Coated Plate is intended for Immunoassay and ELISA. The plate could not be separated, and the frame is clear. Based on the high binding affinity between biotin-streptavidin systems, SP-14 is easier to watch color changes compared with SP-11.
Catalog No. 502330886 Supplier ACROBiosystems Inc Supplier No. SP145PLATES
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This Streptavidin Coated Plate is intended for Immunoassay and ELISA. The plate could not be separated, and the frame is clear. Based on the high binding affinity between biotin-streptavidin systems, SP-14 is easier to watch color changes compared with SP-11.