Learn More
Electron Microscopy Sciences NIGHTSEA™ Stereo Fluorescence Adapter Standard Lamp Base, Cyan Barrier Filter
Catalog No. 501902441
Encompass
The NIGHTSEA™ Stereo Microscope Fluorescence Adapter adapts just about any stereo microscope (dissecting microscope) for fluorescence with no modification to the microscope itself. The modular design lets you easily switch between several different excitation/emission combinations to work with a variety of fluorescent proteins and other fluorophores. There are six different excitation/emission combinations available, plus white light.Excellent for quick screening of your fluorescent genotypes, genotype sorting, fluorescence-aided dissection, injection, or micromanipulation, freeing up your research-grade fluorescence microscopes for more demanding work, and bringing fluorescence into the teaching laboratory. System consists of flexible gooseneck lamp base with power supply, light head, ring adapter for microscope, barrier filter, and filter shield.Color: CyanExcitation: 490-515nmEmission filter: 550 nm longpassFluorophores: YFP, Venus, Lucifer Yellow
Catalog No. 501902441 Supplier Electron Microscopy Sciences Supplier No. SFACY
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The NIGHTSEA™ Stereo Microscope Fluorescence Adapter adapts just about any stereo microscope (dissecting microscope) for fluorescence with no modification to the microscope itself. The modular design lets you easily switch between several different excitation/emission combinations to work with a variety of fluorescent proteins and other fluorophores. There are six different excitation/emission combinations available, plus white light.Excellent for quick screening of your fluorescent genotypes, genotype sorting, fluorescence-aided dissection, injection, or micromanipulation, freeing up your research-grade fluorescence microscopes for more demanding work, and bringing fluorescence into the teaching laboratory. System consists of flexible gooseneck lamp base with power supply, light head, ring adapter for microscope, barrier filter, and filter shield.Color: CyanExcitation: 490-515nmEmission filter: 550 nm longpassFluorophores: YFP, Venus, Lucifer Yellow