Learn More
Eisco™ Streak Plates for Minerals
The off-black, unglazed porcelain of these streak plates show a mineral's color in powder form, or to identify the mineral content of a rock.
Supplier:  Eisco™ ESNGSPBPK12
Catalog No. S012301
Only null left
Description
Description
  • Includes twelve streak plates
  • Used to determine a mineral's color in powder form, or to identify the mineral content of a rock
  • Great for geology and earth science classrooms
  • Streak plates can be cleaned and reused again and again
  • Each plate is made of off-black, unglazed porcelain
  • Streak plates measure 1.9 in. long, 1.9 in. wide, and 0.2 in. high
  • By scraping a specimen across the unglazed, porcelain plates, students learn to identify minerals by examining the powder color of the streak
Specifications
Specifications
Mineral ID Kit
Product Suggestions
Product Suggestions