Learn More
DWK Life Sciences KIMBLE™ Straight Bore Stopcock Plug PTFE, Size 4
Catalog No. K8210010004
Change view
Click to view available options
Material:
PTFE
Catalog No. Material
K8210010004 PTFE
1 options
Catalog No. K8210010004 Supplier DWK Life Sciences Supplier No. 8210010004
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
KIMBLE™ Straight Bore Stopcock Plug PTFE, Size 4 is chemically resistant and self-lubricating, featuring a 1:5 taper for easier use at higher-than-ambient temperatures. Ideal as a replacement plug for KIMBLE™ straight bore stopcocks.
  • Chemically resistant PTFE material, self-lubricating for smooth operation
  • 1:5 taper plug design permits easier use at higher-than-ambient temperatures
  • Bore size: 4 mm; O-ring size: 5 to 613
  • Plug size: 15.2/30 mm
  • Compatible with Kimble straight bore stopcocks (e.g. 821000-0004)
Specifications
Type Straight Bore Stopcock Plug
Plug Bore 4 mm
Material PTFE
For Use With (Application) Stopcocks
Plug Material PTFE