Learn More
Inmark STP-710 - DISPOSABLE 2-PART SECONDARY PRESSURE VESSEL (95 KPA), MEDIUM
Supplier:  Inmark STP710
Encompass
STP-710 DISPOSABLE 2-PART SECONDARY PRESSURE VESSEL (95 KPA), MEDIUM; Inner leak proof polybag 9.25 x 12in.; Tyvek outer envelope 7.5 x 9.5 in.; Maximum Capacity Four 10mL vials or 2in. grid box;, 50/CASE
Catalog No. NC9116473
Only null left