Learn More
Inmark STP-308SYS - Category B / Exempt Frozen Insulated Shipping System (UN 3373)
Catalog No. NC0242762
Encompass
STP-308SYS Category B / Exempt Frozen Insulated Shipping System (UN 3373), Outer Box OD: 12 x10 x 15.75in., Insulated Chest ID: 8.5 x 6.5 x 11.75in., 8/Case
Catalog No. NC0242762 Supplier Inmark Supplier No. STP308SYS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
STP-308SYS Category B / Exempt Frozen Insulated Shipping System (UN 3373), Outer Box OD: 12 x10 x 15.75in., Insulated Chest ID: 8.5 x 6.5 x 11.75in., 8/Case