Learn More
Inmark STP-100 - Category A Ambient Shipping System (UN 2900, UN 2814)
Catalog No. NC9238881
Encompass
STP-100 Outer Box OD 6.5 x 6.5 x 8in., pre-printed with: Class 6 Infectious Substance Label & UN2814 Marking, Reusable polypropylene Secondary Pressure Vessel; Ambient Infectious Substance Shipper, Meets All Requirements of ICAO/IATA Packing Instruction 602, Infectious Substances and All Requirements of National and International Regulations for Surface and Air Transportation, 12/Case
Catalog No. NC9238881 Supplier Inmark Supplier No. STP100C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
STP-100 Outer Box OD 6.5 x 6.5 x 8in., pre-printed with: Class 6 Infectious Substance Label & UN2814 Marking, Reusable polypropylene Secondary Pressure Vessel; Ambient Infectious Substance Shipper, Meets All Requirements of ICAO/IATA Packing Instruction 602, Infectious Substances and All Requirements of National and International Regulations for Surface and Air Transportation, 12/Case