Learn More
Thermo Scientific™ Store™ Informatics Software
Catalog No. S0100015
Encompass
Catalog No. S0100015 Supplier Thermo Scientific™ Supplier No. S0100015
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Store and manage data automatically for rapid, highly scalable, global access to all HCS data.
Using Thermo Scientific™ Store™ Informatics Software, store and manage data automatically for rapid, highly scalable, global access to all HCS data.
Thermo Scientific™ Store™ Image and Database Management Software is a SQL Server or Oracle™database used for storing and managing the images and data automatically transferred from an HCS instrument or imported from another imaging source. It allows for rapid, highly scalable, global access to both data and images. Use the Store Image and Database Management Software after you have collected data and images.
Highlights:
  • Automatic Transfer of Images and Data – Centralized data storage allowing data and image access from multiple locations
  • Accessible and Linked – Data is seamlessly linked to its associated image
  • Visualize – View images and data linked together and easily compare images
  • Export – Built in export formats for software such as Spotfire™, Excel™, and GeneData Screener™ programs
  • HCS Connect – Fully open standard, web services API that provides programmatic access to any data and images being managed by Store software
  • HCS Gateway – Users can then benefit from a single, open standard repository of images and data regardless of source
Specifications
Description Thermo Scientific Store Informatics Software Enterprise Edition