Learn More
Thermo Scientific™ Storage Tube Protection
GREENER_CHOICE
Catalog No. 1535023
Change view
Click to view available options
Product Type:
Cryo Storage Box
Tube Wrap
Vial Holder
Quantity:
100/Pk., 100/Cs.
15/Pk., 300/Cs.
200/Pk., 200/Cs.
350/Pk., 350/Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
1535023 Vial Holder 100/Pk., 100/Cs.
12565177 Tube Wrap 15/Pk., 300/Cs.
12565184 Cryo Storage Box 350/Pk., 350/Cs.
12565185 Cryo Storage Box 200/Pk., 200/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 1535023 Supplier Thermo Scientific™ Supplier No. 50160001
Give your cryogenic storage tubes and vials extra protection, such as when shipping or storing in liquid or vapor phase nitrogen with these storage accessories.
Give your cryogenic storage tubes and vials extra protection when needed with Thermo Scientific™ Storage Tube Protection accessories. For example, safeguard your samples when storing in liquid or vapor phase nitrogen, or when shipping samples. These cryogenic storage accessories, such as tube wraps, sleeves and mini-boxes, have safe handling in mind.
  • If you want to submerge your cryotubes in the liquid phase of nitrogen, seal them first inside Nunc™ Cryoflex™ tubing to prevent liquid nitrogen from infiltrating the tube. Cryoflex™ tubing can be used to crimp and heat shrink an impenetrable safety skin around the tube prior to submersion.
  • Nalgene™ CryoSleeves™ slide over any Nunc or Nalgene Cryocane and prevent tubes from falling out of the cryocane and into the bottom of the liquid nitrogen dewar. Nalgene™ CryoSleeves™ do not prevent infiltration of liquid nitrogen. Cryogenic tubes mounted in the cane must first be sealed inside Cryoflex™ tubing prior to applying the CryoSleeve™ if liquid phase submersion is required.
  • Nunc CryoTube™ miniboxes with a friction-fit lid are ideal for storing small quantities of samples and shipping.Made with durable, high-impact, polystyrene to protect samples.
Specifications
Description Nalgene CryoSleeve, 273 mm, PVC
Quantity 100/Pk., 100/Cs.
Height (Metric) 100 mm
Product Type Vial Holder