Learn More
Chemglass Inc 10mL Flask, Reaction, 14/20 Outer Joint, Valve, Airfree, Schlenk
Catalog No. AF052020
Encompass
10mL Flask, Reaction, 14/20 Outer Joint, Valve, Airfree, Schlenk. Kjeldahl shape facilitates heating and stirring while permitting easy transfer of liquids or solids. Lower portion of flask fits standard heating mantles. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.
Catalog No. AF052020 Supplier Chemglass Inc Supplier No. AF052020
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10mL Flask, Reaction, 14/20 Outer Joint, Valve, Airfree, Schlenk. Kjeldahl shape facilitates heating and stirring while permitting easy transfer of liquids or solids. Lower portion of flask fits standard heating mantles. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.