Learn More
Eppendorf™ Storage Boxes
Allows selection of the optimal box combinations for individual storage demands and provides safe storage for your sample tubes in a space-saving design.
Supplier:  Eppendorf™ 0030140613
 View more versions of this product
Catalog No. 13683536
Only null left
Description
Description
  • Flexible and optimized combination options for the different box formats
  • Retains shape at freezing temperature of -86°C for sample storage or autoclaving for 20 minutes at 121°C for sterilization
  • Reliable individual alpha-numeric laser labeled code assigned to each sample location
  • Minimizes allocation errors
  • Provides high-contrast readability
  • Remains permanently
  • Five additional inscription labels on lid enable user-specific labeling
  • Fits all typical tube formats: cryogenic tubes, microcentrifuge tubes, 15 and 50 mL conical tubes and other laboratory vessels
  • 15 mL and 50 mL conical tube boxes with ventilation slots for an evenly distributed freezing process
  • Prevents tube damage or breakage, which is frequently observed when freezing long tube formats in closed boxes
  • Compatible with common freezer rack systems and the different inner grid variants
Specifications
Chemical Identifiers
Polypropylene
Yes
Storage Box
Specifications
Autoclavable
Polypropylene
Yes
2 pcs.
Storage Box
25 Tubes (5 mL) with Screw Cap
25
5 x 5
3 in.
76.2 mm
Product Suggestions
Product Suggestions