missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Storage Boxes
Allows selection of the optimal box combinations for individual storage demands and provides safe storage for your sample tubes in a space-saving design.
Supplier: Eppendorf™ 0030140532
Description
- Flexible and optimized combination options for the different box formats
- Retains shape at freezing temperature of -86°C for sample storage or autoclaving for 20 minutes at 121°C for sterilization
- Reliable individual alpha-numeric laser labeled code assigned to each sample location
- Minimizes allocation errors
- Provides high-contrast readability
- Remains permanently
- Five additional inscription labels on lid enable user-specific labeling
- Fits all typical tube formats: cryogenic tubes, microcentrifuge tubes, 15 and 50 mL conical tubes and other laboratory vessels
- 15 mL and 50 mL conical tube boxes with ventilation slots for an evenly distributed freezing process
- Prevents tube damage or breakage, which is frequently observed when freezing long tube formats in closed boxes
- Compatible with common freezer rack systems and the different inner grid variants
Chemical Identifiers
| Polypropylene | |
| Yes | |
| Storage Box |
Specifications
| Autoclavable | |
| Polypropylene | |
| Yes | |
| 4 pcs. | |
| Storage Box |
| 25 Tubes (5 mL) | |
| 25 | |
| 5 x 5 | |
| 2.5 in. | |
| 63.5 mm |