Learn More
Microsolv Technology Storage Box for 4ml Autosampler Vials. Red, chemical resistant polypropylene holds 49 vials. Complete with translucent plastic lid. 1 EA. DFS Item
Supplier:  Microsolv Technology Corporation 9504SB03
Excellent for standing position on the lab bench or during transport and ideal for space saving storage in places such as refrigerators. Lid prevents condensation on the caps minimizing contamination during storage. Usable temperature range -80C to +100C. Alpha numeric indexing coded on bottom of wells and along the sides of the box for clear sample identification. Box outer dimensions are 130mm wide by 130mm deep by 52mm high. Box and lid are Autoclavable.
Catalog No. 50-227-1360
Only null left
Explore available promotions