missing translation for 'onlineSavingsMsg'
Learn More

Stoddard Solvent

Catalog No. AC419730010
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC419730010 1 kg
AC419730050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC419730010 Supplier Thermo Scientific Chemicals Supplier No. 419730010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 8052-41-3 | C8H15BrO2 | 223.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8052-41-3
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00132767
InChI Key IMSPZLCTPSXORJ-UHFFFAOYSA-N
Synonym 6-bromo-1-hexanol acetate, 1-acetoxy-6-bromohexane, stoddard solvent, 1-hexanol,6-bromo-,acetate, neo-clear r xylene substitute, w/w benzene, acetic acid 6-bromohexyl ester, 6-acetoxy-1-bromohexane
PubChem CID 11806291
IUPAC Name 6-bromohexyl acetate
SMILES CC(=O)OCCCCCCBr

Specifications

Melting Point -40.0°C
Density 0.7870g/mL
Quantity 1 kg
Flash Point >38°C
Packaging Glass Bottle
Physical Form Solvent
Specific Gravity 0.787
UN Number 1268
Boiling Point 149.0°C to 213.0°C
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Stoddard solvent
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement May be fatal if swallowed and enters airways. Toxic to aquatic life with long lasting effects. Flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. Do not get in eyes, on skin, or on clothing. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting.
EINECSNumber 232-489-3
RTECSNumber WJ8925000
TSCA TSCA

RUO – Research Use Only