Learn More
Chemglass Inc Stirrer Blade, PTFE, 10mm, for 500mL to 1000mL Flasks, 19mm Width x 75mm Length, 24/40 minimum joint clearance
Catalog No. CG208003
Encompass
Stirrer Blade, PTFE, 10mm, for 500mL to 1000mL Flasks, 19mm Width x 75mm Length, 24/40 minimum joint clearance. Stirrer blade made of 1/8" thick solid PTFE for use with both Chem-Stir™ and button style stirring shafts having a 10mm OD.
Catalog No. CG208003 Supplier Chemglass Inc Supplier No. CG208003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Stirrer Blade, PTFE, 10mm, for 500mL to 1000mL Flasks, 19mm Width x 75mm Length, 24/40 minimum joint clearance. Stirrer blade made of 1/8" thick solid PTFE for use with both Chem-Stir™ and button style stirring shafts having a 10mm OD.