missing translation for 'onlineSavingsMsg'
Learn More

Stiripentol 98.0+%, TCI America™

Catalog No. S09055G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09055G 5 g
S09051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09055G Supplier TCI America Supplier No. S09055G

Chemical Identifiers

CAS 49763-96-4
Molecular Formula C14H18O3
Molecular Weight (g/mol) 234.295
MDL Number MFCD00869310
InChI Key IBLNKMRFIPWSOY-FNORWQNLSA-N
Synonym 4,4-Dimethyl-1-(3,4-methylenedioxyphenyl)-1-penten-3-ol, 1-(1,3-Benzodioxol-5-yl)-4,4-dimethyl-1-penten-3-ol
PubChem CID 5311454
IUPAC Name (E)-1-(1,3-benzodioxol-5-yl)-4,4-dimethylpent-1-en-3-ol
SMILES CC(C)(C)C(C=CC1=CC2=C(C=C1)OCO2)O

Specifications

Melting Point 77°C
Color White
Quantity 5 g
Formula Weight 234.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Stiripentol
RTECSNumber SB3393000
TSCA No
Recommended Storage Freezer