missing translation for 'onlineSavingsMsg'
Learn More

Stilbene Yellow, Spectrum™ Chemical

Catalog No. 18614092
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614092 25 g Amber Glass Bottle
18614091 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614092 Supplier Spectrum Chemical Mfg Cor Supplier No. ST12025GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1325-37-7

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 g
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Stilbene Yellow