missing translation for 'onlineSavingsMsg'
Learn More

Stigmasterol 90.0+%, TCI America™

Catalog No. p-7135802
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00881G 1 g
S00885G 5 g
S008825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S00881G Supplier TCI America Supplier No. S00881G

Chemical Identifiers

CAS 83-48-7
Molecular Formula C29H48O
Molecular Weight (g/mol) 412.70
MDL Number MFCD00003630
InChI Key HCXVJBMSMIARIN-PHZDYDNGSA-N
Synonym stigmasterol, stigmasterin, beta-stigmasterol, stigmasta-5,22-dien-3beta-ol, 24s-5,22-stigmastadien-3beta-ol, unii-99wuk5d0y8, stigmasta-5,22e-dien-3beta-ol, stigmasta-5,22-dien-3-beta-ol, 3beta,22e-stigmasta-5,22-dien-3-ol, stigmasta-5,22-dien-3-ol, 3b,22e
PubChem CID 5280794
ChEBI CHEBI:28824
IUPAC Name (1R,3aS,3bS,7S,9aR,9bS,11aR)-1-[(2R,3E,5S)-5-ethyl-6-methylhept-3-en-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
SMILES CC[C@H](\C=C\[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C

Specifications

Melting Point 167°C
Color White
Quantity 1 g
Formula Weight 412.70
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Stigmasterol
EINECSNumber (9)-1840
RTECSNumber WJ2447500
TSCA No