missing translation for 'onlineSavingsMsg'
Learn More

Stigmasterol, 95%, Thermo Scientific Chemicals

Catalog No. p-193574
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132760050 5 g Glass bottle
AC132760250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC132760050 Supplier Thermo Scientific Chemicals Supplier No. 132760050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83-48-7
Molecular Formula C29H48O
Molecular Weight (g/mol) 412.70
MDL Number MFCD00003630
InChI Key HCXVJBMSMIARIN-PHZDYDNGSA-N
Synonym stigmasterol, stigmasterin, beta-stigmasterol, stigmasta-5,22-dien-3beta-ol, 24s-5,22-stigmastadien-3beta-ol, unii-99wuk5d0y8, stigmasta-5,22e-dien-3beta-ol, stigmasta-5,22-dien-3-beta-ol, 3beta,22e-stigmasta-5,22-dien-3-ol, stigmasta-5,22-dien-3-ol, 3b,22e
PubChem CID 5280794
ChEBI CHEBI:28824
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-17-[(E,2R,5S)-5-ethyl-6-methylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC[C@H](\C=C\[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C

Specifications

Melting Point 161.0°C to 170.0°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 8944
Specific Rotation Condition - 50.00 (20.00°C c=2,CHCl3)
Specific Rotation − 50.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 412.7
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Stigmasterol, 95%
Show More Show Less
EINECSNumber 201-482-7

RUO – Research Use Only