missing translation for 'onlineSavingsMsg'
Learn More

Stevioside 85.0+%, TCI America™

Catalog No. S059425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S059425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S059425G Supplier TCI America Supplier No. S059425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57817-89-7
Molecular Formula C38H60O18
Molecular Weight (g/mol) 804.88
MDL Number MFCD00079561
InChI Key UEDUENGHJMELGK-PEKONUEISA-N
Synonym stevioside
PubChem CID 134129657
SMILES CC12CCCC(C1CCC34C2CCC(C3)(C(=C)C4)OC5C(C(C(C(O5)CO)O)O)OC6C(C(C(C(O6)CO)O)O)O)(C)C(=O)OC7C(C(C(C(O7)CO)O)O)O

Specifications

Melting Point 236°C
Color White-Yellow
Quantity 25 g
Formula Weight 804.88
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Stevioside
RTECSNumber NZ8175000
TSCA No