Learn More
United Scientific™ Stethoscope
Catalog No. S99444
Change view
Click to view available options
Product Type:
Bowles Type Stethoscope
Catalog No. Product Type
S99444 Bowles Type Stethoscope
1 options
This item is intended for educational purposes only.
Catalog No. S99444 Supplier United Scientific™ Supplier No. STHB01
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This item is intended for educational purposes only.
Listen to vital signs with this Stethoscope.
  • Ideal for the classroom, especially in the study of respiration and circulation
  • Teaching stethoscope has 3.8 cm Bowles chestpiece and two sets of binaurals joined together so the teacher and student can listen simultaneously
  • Bowles-type stethoscope has adjustable, chrome-plated binaurals and PVC Y tubing
Specifications
Product Type Bowles Type Stethoscope
Color Black
Includes Adjustable chrome plated binaurals, PVC Y-tubing