missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sterox SE, 1% (w/v) Aqueous Solution, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Sterox SE |
| Color | Yellow |
| CAS | 9004-83-5, 7732-18-5 |
| CAS Min % | 97.0, 0.97 |
| CAS Max % | 100.0, 1.01 |
| Packaging | Poly natural |
| Solvent or Matrix | Aqueous Solution |
| Quantity | 500 mL |
| Physical Form | Soapy Solution |
| Concentration or Composition Notes | 1% (w/v) |