Learn More
United Scientific™ SteriTrue™ Sterile Sampling Scoops, Polystyrene
SteriTrue Scoops are individually packed and are sterile.
Supplier:  United Scientific™ UNST102
 View more versions of this product
Catalog No. 02-923-194
Only null left
Explore available promotions
Alert:
Prop 65
Description
Description
United Scientific™ SteriTrue™ PS sterile sampling scoops are unique designs developed with end-users! The polystyrene scoops are ergonomically designed for long periods of use, they can hold stated volumes, and each scoop has a flat bottom surface for stability during weighing. The most unique feature of the scoops is the long handle: the cross ribs add structural support to help prevent breakage, and the bottom curved design fits comfortably in the palm of the hand.
  • Ergonomically designed with voice-of-customer input
  • Flat bottom surface for stability during weighing
  • Gamma sterilized, individually packaged
  • Lot stamped for traceability
  • PS material meets FDA guidelines and is safe for food, pharma, and other uses
  • Available in Pack and Case quantities
Specifications
Specifications
100/Pk.
250 mL
Polystyrene
Not Autoclavable
Scoop
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Cancer and Reproductive Harm