Learn More
United Scientific™ SteriTrue™ Sterile Sampling Scoops, Metal Detectable Polystyrene
SteriTrue Scoops are metal detectable for easy X-Ray identification and are individually packed and are sterile.
Supplier:  United Scientific™ UNST104
 View more versions of this product
Catalog No. 02-923-198
Only null left
Explore available promotions
Alert:
Prop 65
Description
Description
The United Scientific™ line of sampling tools now includes SteriTrue™ metal detectable polystyrene sterile scoops. These incorporate the ergonomic design of our standard PS line and are permeated with a semi-ferrous additive. This allows the scoops to be easily identified by metal detection and X-ray equipment. The scoops are ergonomically designed for long periods of use, they can hold stated volumes, and each scoop has a flat bottom surface making it easy for weighing samples.
  • Metal detectable PS resin
  • Ergonomically designed with voice-of-customer input
  • Flat bottom surface for stability during weighing
  • Gamma sterilized, individually packaged
  • Lot stamped for traceability
  • PS material meets FDA guidelines and is safe for food, pharma, and other uses
Specifications
Specifications
100/Pk.
125 mL
Polystyrene Resin
Not Autoclavable
Scoop
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Cancer and Reproductive Harm