Learn More
Sartorius Sterisart ™ Universal Pump, Gen 4
The Sterisart™ Universal Gen 4 pump is engineered for seamless operation within barrier technology environments, offering precise and contamination-free transfer of samples and media into Sterisart™ canisters for sterility testing.
Supplier:  Sartorius 16421A
 View more versions of this product
Catalog No. 14222733
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Universal sterility testing pump designed for benchtop use or for integration in isolators
  • Low profile and compact footprint
  • Flexible, modular and open design
  • 316 L mirror finish stainless steel
  • Sleek design for impeccable hygiene: easy disassembly and smooth contours
  • Real-Time pressure detection in the Sterility testing canisters
  • 7 in. high performance touch screen display with integrated barcode scanner
  • Built for 21 CFR Part 11 and EU GMP Annex 11: Advanced software for sterility testing featuring user access control, audit trail, secure data handling, electronic signatures, and reports
Specifications
Chemical Identifiers
Sterility Testing Pump
Specifications
21 CFR Part 11
Yes
Sterility Testing Pump
Product Suggestions
Product Suggestions