missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ Sterilized uTIP™ Universal Compatible TipEject™ Reload

Catalog No. 12111422
Change view
Click to view available options
Volume (Metric):
10 μL
Color Code:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Code
12111422 10 μL Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12111422 Supplier BIOTIX™ Supplier No. 63305954

uTIP™ Universal Compatible TipEject™ self-aligning reload solution allows you to easily reload your racks without coming into contact with the tips. The rigid 100% recycled plastic covering shields unused tips from contamination for later use.

  • Tip Eject saves space on lab bench while providing an easy manual ejection system for racking tips
  • Universal compatible pipette tips
  • 100% recycled packaging material
  • Easy-to-use fast reload system
  • Reduces plastic waste by 30% when compared to racked tips
  • Vertically stacked snapcards take up less space on your lab bench
  • FlexFit™ technology creates a seal with less insertion forces than that of competitors
  • X-Resin™ maximizes sample recovery and accuracy by reducing surface to sample binding
  • Blade™ reduces the frequency of hanging droplet formation, improving precision and accuracy

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Rack Color Clear
Autoclavable Autoclavable
Clean Claims RNase, DNase, ATP, pyrogen, endotoxin and DNA Free
Sterilization Method E-Beam
Features FlexFit, X-Resin, Blade
Format Refill
Tip Style Extended Length, Ergonomic, Low Retention
Product Type Pipette Tips
Color Code Red
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line uTip
Material Polypropylene
Fit for Purpose Certified Quality
Graduations 2 μL, 10 μL
Show More Show Less