missing translation for 'onlineSavingsMsg'
Learn More

BD Sterility Bottles

Catalog No. df9065780
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
200 mL
300 mL
500 mL
Color:
Amber
Beige
Clear
Light Beige
Light Tan
Light Yellow
Yellow
Media Type:
Fluid A
Fluid D
Fluid Thioglycollate Medium
Plate Count Agar
Potato Dextrose Agar
R2A Agar
Trypticase Soy Broth
Trypticase™ Soy Broth
Catalog No. Quantity Color Media Type
DF9065780 300 mL Light Yellow Fluid A
B99102 500 mL Yellow Plate Count Agar
DF9066780 300 mL Clear Fluid D
B299906 500 mL Light Tan Potato Dextrose Agar
B99107 100 mL Beige Trypticase Soy Broth
BB99417 100 mL Light Beige Fluid Thioglycollate Medium
L99112 500 mL Light Beige Fluid Thioglycollate Medium
22227315 500 mL Beige R2A Agar
B99108 100 mL Amber Fluid Thioglycollate Medium
DF908274 100 mL Clear Fluid A
B99113 500 mL Light Beige Trypticase™ Soy Broth
B99094 200 mL Beige Plate Count Agar
DF908374 100 mL Clear Fluid D
13 options

Catalog No. DF9065780

Supplier: BD 290652

Only null left

Includes

Certificate of Analysis

Glass sterility bottles available with a variety of media

  • Transparent labels for easy viewing and identification of medium
  • Labels are laminated acetate to resist alcohol or chlorine treatment for cleanroom use
  • Prepared bottles for sterility testing and microbial limits

Wide-Mouth Bottles

  • Openings are 48mm in diameter to accommodate bulky samples

Order Info

Supplied in packs of 10 bottles

Compliance

Conform to harmonized USP requirements.

Specifications

Cap Type Screw
Certifications/Compliance USP
Color Light Yellow
Form Liquid
Manufacturing Origin United States
Media Type Fluid A
Fill Volume 300 mL
Formulation 0.1% Peptic digest of animal tissue
For Use With (Application) Sterility testing and microbial limits
Includes Certificate of Analysis
Packaging Type Bottle
Quantity 300 mL
Show More Show Less