missing translation for 'onlineSavingsMsg'
Learn More

BD Sterility Bottles

Glass sterility bottles available with a variety of media

Supplier:  BD 299094

 View more versions of this product

Catalog No. B99094

Only null left
Explore available promotions

Includes: Certificate of Analysis

Description

Description

  • Transparent labels for easy viewing and identification of medium
  • Labels are laminated acetate to resist alcohol or chlorine treatment for cleanroom use
  • Prepared bottles for sterility testing and microbial limits

Wide-Mouth Bottles

  • Openings are 48mm in diameter to accommodate bulky samples

Order Info

Supplied in packs of 10 bottles

Compliance

Conform to harmonized USP requirements.

Specifications

Specifications

Screw
Beige
Liquid
Plate Count Agar
Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 1g/L, Agar 15g/L
Certificate of Analysis
200 mL
USP
15°C to 30°C
United States
200 mL
Obtaining microbial plate counts from Milk and dairy products, foods, Water and other materials of sanitary importance
Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions