Learn More
Bel-Art™ Sterileware Triangular Scraper
Sterile, plastic scrapers are perfect for cutting, cleaning and general scraping
Supplier:  Bel-Art™ F368210110
Catalog No. 01184165
Only null left
Explore available promotions
Description
Description
  • Ideal for use in the manufacture and quality assurance of creams, ointments and pastes
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) scrapers heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
100 Pk.
Spatula End, Sharpened End
11 cm
22.5 cm
Not Autoclavable
Scraper
HDPE
Product Suggestions
Product Suggestions