missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Sterile Vented PETG Erlenmeyer Flask with Flat Bottom
Description
- Erlenmeyer Flask - Sterile PETG flask with vented caps.
- Flat Bottom Design - Provides stability for gentle stirring, media preparation, and general chemical reactions.
- Vented Cap for Gas Exchange - Allows airflow while preventing contamination, ideal for biological cultures and chemical reactions.
- Gamma Sterilized and Lab complaint - Manufactured to USP Class VI standards, ISO 11137 sterilization, and FDA 21 CFR 177.1630 food contact compliance.
- Wide Scientific Applications - Suitable for biopharmaceutical processing, cell culture, and chemical storage in laboratory settings.
- The flat bottom design provides excellent stability for routine laboratory procedures.
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Material | PETG |
| No. per Pack | 6/Pk. |
| Product Type | Erlenmeyer Flask |
| Autoclavable | Not Autoclavable |
| Disposable | Single-use |
| Graduated | Yes |
| Mouth Style | Narrow |
| Sterility | Sterile |
| Bottom Shape | Flat |