missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Sterile Universal Filter Tips, Extended Length

Catalog No. 22171784
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 1000 μL
Length (Metric):
45 mm
105 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Length (Metric)
22171784 1 to 1000 μL 105 mm
22171778 0.1 to 10 μL 45 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22171784 Supplier Globe Scientific Supplier No. 150938
Only null left

Manufactured in a Class 100,000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems.

  • Manufactured from clear polypropylene (PP)
  • Polypropylene (PP) filters protect samples and pipettes from contamination
  • Dnase-free, Rnase-free and non-pyrogenic
  • Manufactured in a Class 100,000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems
  • Sterilized by gamma irradiation SAL10-6 (ISO11137)
  • Racks are printed with lot number for quality traceability

Specifications

Volume (Metric) 1 to 1000 μL
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Length (Metric) 105 mm
Clean Claims Rnase, Dnase, and Pyrogen Free
Sterilization Method Gamma Irradiation
Format Rack
Tip Style Extended Length
Packaging 96 Tips/Rack, 10 Racks/Pk, 2 Boxes/Cs.
For Use With (Equipment) Universal Fit
Graduated Yes
Show More Show Less